

(±)-1-Ethoxy-2-propanol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1569-02-4
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00067050
:104.15 g/mol
Boiling Pt:132 °C (1013 hPa)
Melting Pt:–59 °C
Density:0,9028 g/cm³ (20 °C)
Flash Pt:38°C
EINECS:216-374-5
UN:1987
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together