

Dibenzyl ether ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:103-50-4
:
Formula:C₆H₅CH₂OCH₂C₆H₅
MDL Number:MFCD00004780
:198.26 g/mol
Merck Index:14,01132
Boiling Pt:298 °C (1013 hPa)
Density:1,0424 g/cm³ (20 °C)
Flash Pt:135°C
EINECS:203-118-2
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together