
N-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)
: TCI
:94-68-8
:
:C₉H₁₃N
:MFCD00026695
:135.21 g/mol
:214°C
:–15 °C
:0,95 g/cm³ (20 °C)
:98°C
:Ambient
:202-354-3
:2754
:6.1,II
:GHS06
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together