
2,4-Diamino-6-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
TCIAD1321-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:very faint turbidity
:
:126.12 g/mol
:14,02981
:373 °C (1013 hPa)
:285…286 °C
:200-254-4
:GHS07
:56-06-4
:MFCD08528919
:C₄H₆N₄O
:MFCD00006098