


1,5-Diaminonaphthalene ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2243-62-1
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004029
:158.2 g/mol
Boiling Pt:381 °C (1013 hPa)
Melting Pt:188 °C
Density:1,4 g/cm³ (20 °C)
EINECS:218-817-8
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Amber to Dark purple powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:187.0 to 194.0 deg-C
Specification Test Results
Frequently Bought Together