


Cesium iodide ≥99.0% (by titrimetric analysis)
Catalog # TCIAC2205-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity:min. 99.0 %
Specification Test Results
About this item
:
:259.81 g/mol
Merck Index:14,02014
Boiling Pt:1280 °C (1013 hPa)
Melting Pt:621°C
Density:3,21 g/cm³ (20 °C)
EINECS:232-145-2
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
CAS Number:7789-17-5
Formula:CsI
MDL Number:MFCD00010962