Order Entry
Netherlands
ContactUsLinkComponent
 
 
2'-(Trifluoromethyl)acetophenone ≥98.0% (by GC)
  TCIAT1899-5G
 :  TCI
 :  
undefined
 
2'-(Trifluoromethyl)acetophenone ≥98.0% (by GC)
  TCIAT1899-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :188.15 g/mol
 :163 °C (1013 hPa)
 :15…17 °C
 :1,25 g/cm³ (20 °C)
 :84°C
 :241-434-2
 :GHS07
 :17408-14-9
 :C₉H₇F₃O
 :MFCD00000378