

α,α,α-Trifluoro-2-tolualdehyde ≥97.0% (by GC)
Catalog # TCIAT1281-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Purity (GC):min. 97.0 %
- Appearance:Colorless to Light yellow clear liquid
Specification Test Results
About this item
:
:174.12 g/mol
Boiling Pt:70°C
Melting Pt:–40 °C
Density:1,322 g/cm³ (20 °C)
Flash Pt:52°C
EINECS:207-185-9
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:447-61-0
Formula:C₈H₅F₃O
MDL Number:MFCD00003337