
1,1'-Thiocarbonyldiimidazole ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6160-65-2
:
Formula:C₇H₆N₄S
MDL Number:MFCD00005289
:178.22 g/mol
Melting Pt:105…106 °C
EINECS:228-183-4
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:97.0 to 106.0 deg-C
Specification Test Results
Frequently Bought Together