
Potassium hydrogen phthalate ≥99.8% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:877-24-7
:
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070
:204.22 g/mol
Merck Index:14,07612
Melting Pt:295…300 °C
Density:1,6362 g/cm³ (20 °C)
EINECS:212-889-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.5 area%
- Purity (Neutralization titration):99.8 to 100.2 %
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together