

Caffeic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:331-39-5
:
Formula:C₉H₈O₄
MDL Number:MFCD00004392
:180.16 g/mol
Merck Index:14,01635
Boiling Pt:366 °C (1013 hPa)
Melting Pt:234…237 °C
EINECS:206-361-2
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together