Order Entry
Netherlands
ContactUsLinkComponent
trans-2-Methoxycinnamic acid ≥99.0% (by titrimetric analysis)
Irritant
trans-2-Methoxycinnamic acid ≥99.0% (by titrimetric analysis)
Catalog # TCIAM0449-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
trans-2-Methoxycinnamic acid ≥99.0% (by titrimetric analysis)
Catalog # TCIAM0449-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    184.0 to 187.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :178.19 g/mol
EINECS:213-785-1
 :GHS07
CAS Number:1011-54-7
 :6099-03-2
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00064238