
4-Nitrosalicylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:619-19-2
:
Formula:C₇H₅NO₅
MDL Number:MFCD00071505
:183.12 g/mol
EINECS:210-584-0
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together