
3-Methoxyanthranilic acid ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3177-80-8
:
Formula:C₈H₉NO₃
MDL Number:MFCD00075178
:167.16 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:169.0 to 173.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together