DL-Histidine ≥98.0% (by HPLC, titration analysis)
: TCI
:4998-57-6
:
:C₆H₉N₃O₂
:MFCD00005208
:155.16 g/mol
:14,04720
:273 °C
:225-660-9
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Frequently Bought Together