Potassium sorbate ≥99.0% (by titrimetric analysis)
: TCI
:24634-61-5
:590-00-1
:CH₃CH=CHCH=CHCOOK
:MFCD00016546
:150.22 g/mol
:14,07671
:270 °C
:1,36 g/cm³ (20 °C)
:246-376-1
:GHS07
:Ambient
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Water:almost transparency
Frequently Bought Together