

1,3-Dimethylbarbituric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:769-42-6
:
Formula:C₆H₈N₂O₃
MDL Number:MFCD00006675
:156.14 g/mol
EINECS:212-211-7
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:122.0 to 125.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together