
2'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:118-93-4
:MFCD00002219
Formula:C₈H₈O₂
:136.15 g/mol
Boiling Pt:218°C
Melting Pt:6°C
Density:1,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-288-0
:GHS07
Specification Test Results
- Appearance:Colorless to Orange to Green clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together