

4,4'-Bis(diethylamino)benzophenone ≥92.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:90-93-7
:
Formula:C₂₁H₂₈N₂O
MDL Number:MFCD00009044
:324.47 g/mol
Boiling Pt:420 °C (1013 hPa)
Melting Pt:96.5 °C
Storage Temperature:Ambient
EINECS:202-025-4
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Blue to Dark blue powder to crystal
- Purity (HPLC):min. 92.0 area%
- Purity (Nonaqueous Titration):min. 92.0 %
Specification Test Results
Frequently Bought Together