9-Fluorenone ≥98.0% (by GC)
: TCI
:486-25-9
:
:C₁₃H₈O
:MFCD00001141
:180.21 g/mol
:342°C
:81…83 °C
:0,9 g/cm³ (20 °C)
:Ambient
:207-630-7
- Appearance:Light yellow to Yellow to Orange flake
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 86.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together