

4-Cyanobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6068-72-0
:
Formula:C₈H₄ClNO
MDL Number:MFCD00001822
:165.58 g/mol
Boiling Pt:110°C
Melting Pt:78.5 °C
EINECS:228-005-5
UN:3261
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:68.0 to 71.0 deg-C
Specification Test Results
Frequently Bought Together