
2-Chlorothioanisole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17733-22-1
:
Formula:C₇H₇ClS
MDL Number:MFCD00040124
Boiling Pt:227°C
ADR:9,
:158.65 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together