Order Entry
Netherlands
ContactUsLinkComponent
Chloral hydrate ≥99.0% (by titrimetric analysis)
 
Chloral hydrate ≥99.0% (by titrimetric analysis)
  TCIAC0073-500G
 :  TCI
 :  
undefined
 
Chloral hydrate ≥99.0% (by titrimetric analysis)
  TCIAC0073-500G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Appearance:
    White or Colorless powder to crystal
  • Purity (Neutralization back titration):
    min. 99.0 %
  • Solubility in EtOH (95vol%):
    almost transparency

 

 

 :
 :165.4 g/mol
 :14,02071
 :97,5 °C (1013 hPa)
 :52 °C
 :1,901 g/cm³ (20 °C)
 :Ambient
 :206-117-5
 :2811
 :6.1,III
 :GHS06
 :302-17-0
 :Cl₃CCH(OH)₂
 :MFCD00044479