
Choline chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:67-48-1
:
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721
:139.62 g/mol
Merck Index:14,02206
Melting Pt:302°C
Density:~ 1,205 g/cm³ (20 °C)
EINECS:200-655-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (after drying)
- Purity (Argentmetric Titration):min. 98.0 % (after drying)
- Solubility in Water:almost transparency
- Drying loss:max. 1.0 %
Specification Test Results
Frequently Bought Together