Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,3-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
  TCIAD2924-5G
 :  TCI
 :  
undefined
 
2,3-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
  TCIAD2924-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :140°C
 :1,321 g/cm³ (20 °C)
 :105°C
 :8,III
 :176.04 g/mol
 :2735
 :GHS05
 :39226-95-4
 :C₇H₇Cl₂N
 :MFCD00047927