Order Entry
Netherlands
ContactUsLinkComponent
1,8-Dichloro-9,10-anthraquinone ≥95.0% (by GC)
Irritant
1,8-Dichloro-9,10-anthraquinone ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
1,8-Dichloro-9,10-anthraquinone ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Green to Brown powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    201.0 to 206.0 deg-C

Specification Test Results

Frequently Bought Together