

2,3-Dichloropyrazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4858-85-9
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00040964
:148.98 g/mol
EINECS:225-460-1
:GHS07
:GHS08
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together