Order Entry
Netherlands
ContactUsLinkComponent
Chlorodiphenylphosphine ≥97.0% (by titrimetric analysis)
Corrosive
Chlorodiphenylphosphine ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Chlorodiphenylphosphine ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (Argentmetric Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together