
2,6-Dibromo-4-nitrophenol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0197-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:296.9 g/mol
Melting Pt:148.5 °C
EINECS:202-744-3
:GHS07
CAS Number:99-28-5
Formula:C₆H₃Br₂NO₃
MDL Number:MFCD00007334