Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
 
1-Bromo-2-fluoro-5-nitrobenzene ≥95.0% (by GC)
 :  TCI
 
undefined
 
1-Bromo-2-fluoro-5-nitrobenzene ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow to Light orange powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    55.0 to 59.0 deg-C

 

Frequently Bought Together