Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-3-fluorobenzoic acid ≥96.0% (by GC, titration analysis)
Irritant
2-Bromo-3-fluorobenzoic acid ≥96.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
2-Bromo-3-fluorobenzoic acid ≥96.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Neutralization titration):
    min. 96.0 %
  • Melting point:
    158.0 to 163.0 deg-C

Specification Test Results

Frequently Bought Together