
1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:328-70-1
:
Formula:C₈H₃BrF₆
MDL Number:MFCD00000381
:293.01 g/mol
Boiling Pt:151°C
Melting Pt:–16 °C
Density:1,7048 g/cm³ (20 °C)
Flash Pt:110°C
EINECS:206-334-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together