Weighing bottles, glass
Made of borosilicate glass.
- Excellent chemical resistance
- Excellent temperature resistance
With interchangeable lid and frosted label.
Frequently Bought Together
Made of borosilicate glass.
With interchangeable lid and frosted label.