
Ethyl-N-hydroxyacetimidate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10576-12-2
:
Formula:C₄H₉NO₂
MDL Number:MFCD00002114
:103.12 g/mol
Boiling Pt:55…58 °C (6 mmHg)
Melting Pt:23…25 °C
Flash Pt:77 °C (171 °F)
EINECS:234-165-7
:GHS07
Related Information
Frequently Bought Together