Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethoxycyclohexanone
2-Ethoxycyclohexanone ≥98%
2-Ethoxycyclohexanone ≥98%
2-Ethoxycyclohexanone ≥98%
H27613.06
:
Thermo Fisher Scientific
:
2-Ethoxycyclohexanone ≥98%
H27613.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
:
33371-97-0
:
:
C₈H₁₄O₂
:
142.20 g/mol
:
85 °C (15 mmHg)
:
1.002
:
54 °C (129 °F)
:
Ambient
:
1224
:
3,III
:
GHS02
:
MFCD00015377