Order Entry
Netherlands
ContactUsLinkComponent
2,4,6-Trichloroanisole-[D5], Supelco®
Irritant
2,4,6-Trichloroanisole-[D5], Supelco®
Catalog # 34023-50MG-R
Supplier:  Merck
CAS Number:  
undefined
Irritant
2,4,6-Trichloroanisole-[D5], Supelco®
Catalog # 34023-50MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Ambient
CAS Number:352439-08-8
Formula:C₇D₅Cl₃O