Herbicide referentiestandaard, 4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB) 100 µg/ml in acetonitrile
EHERXA11980000AL
:
- Verpakking:Glass bottle
- VPE:1 mL
- CAS No.:94-82-6
- Omschrijving:4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB) 100 µg/ml in acetonitrile
- Formule:C₁₀H₁₀Cl₂O₃
- Omschrijving:4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB)