Luminol (3-Aminophthalhydrazide) 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:521-31-3
:
Formula:C₈H₇N₃O₂
MDL Number:MFCD00006890
Melting Pt:319…320 °C
:177.16 g/mol
EINECS:208-309-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to yellow-green to beige Powder and/or chunks
- Infrared spectrum:Conforms
- Solubility:(2 % in 1 N NaOH) Clear yellow to yellow-green
- Titration excess NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together