Order Entry
Netherlands
ContactUsLinkComponent
(2S,3S)-(+)-Diethyl-2,3-O-benzylidenetartrate for synthesis
(2S,3S)-(+)-Diethyl-2,3-O-benzylidenetartrate for synthesis
Catalog # 8.14041.0005
Supplier:  Merck
CAS Number:  
(2S,3S)-(+)-Diethyl-2,3-O-benzylidenetartrate for synthesis
Catalog # 8.14041.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
CAS Number:141042-56-0
Formula:C₁₅H₁₈O₆