DL-Carnitine hydrochloride
Catalog # ROTH7212.1
Leverancier: Roth Carl
CAS-nummer:
Specificaties
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specificaties
Over dit artikel
:
:197.66 g/mol
EINECS:207-309-1
CAS-nummer:461-05-2
Formule:C₇H₁₆ClNO₃
MDL-nummer:MFCD00011904