

DL-Carnitine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:461-05-2
:
Formula:C₇H₁₆ClNO₃
MDL Number:MFCD00011904
:197.66 g/mol
Melting Pt:190…205 °C
EINECS:207-309-1
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:194 to 200 °C
- Specific optical rotation:-1° to +1° (20 °C, 589 nm) (c=1, H2O)
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together