
(4-Methoxybenzyl)triphenylphosphonium chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3462-97-3
:
Formula:C₂₆H₂₄ClOP
MDL Number:MFCD00031572
:418.91 g/mol
Melting Pt:235…239 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together