
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1253-47-0
:
Formula:C₂₇H₂₄ClO₂P
MDL Number:MFCD02683069
:446.91 g/mol
Melting Pt:230…235 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together