Order Entry
Netherlands
ContactUsLinkComponent
(4-Cyanobenzyl)triphenylphosphonium chloride for synthesis
(4-Cyanobenzyl)triphenylphosphonium chloride for synthesis
Catalog # 8.18511.0005
Supplier:  Merck
CAS Number:  
(4-Cyanobenzyl)triphenylphosphonium chloride for synthesis
Catalog # 8.18511.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    326 - 338 °C
  • Molar Mass:
    413.88 g/mol
  • Synonyms:
    Triphenyl-4-cyanobenzylphosphonium chloride
  • Hill Formula:
    C26H21ClNP

Specification Test Results

About this item

 :
 :413.89 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:20430-33-5
Formula:C₂₆H₂₁ClNP
MDL Number:MFCD00011752