Ga naar Rubriek
Ga naar Hoofdinhoud
Ga naar Voettekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, Nicotinamide 10 µg/ml in methanol
Organic reference standard, Nicotinamide 10 µg/ml in methanol
Organic reference standard, Nicotinamide 10 µg/ml in methanol
PROONE9289
:
LGC Standards PROMOCHEM
:
Organic reference standard, Nicotinamide 10 µg/ml in methanol
PROONE9289
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
98-92-0
Description:
Nicotinamide 10 µg/ml in methanol
Formula:
C₆H₆N₂O
Description:
Nicotinamide