Order Entry
Netherlands
ContactUsLinkComponent
Anthracene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Anthracene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalogus # EHERL20520100CY
Supplier:  EHRENSTORFER
CAS Number:  
Anthracene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalogus # EHERL20520100CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    1719-06-8
  • Description:
    Anthracene-[D10] 10 µg/ml in cyclohexane
  • Formula:
    C₁₄D₁₀
  • Description:
    Anthracene-[D10]

Specifications

About this item