
1-Chloro-2-fluoro-4-nitrobenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:350-31-2
:
Formula:C₆H₃ClFNO₂
MDL Number:MFCD00215833
:175.55 g/mol
Boiling Pt:116°C
EINECS:206-500-7
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:58.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together