Order Entry
Netherlands
ContactUsLinkComponent
Trichloroacetic acid (TCA) 500 g/l in aqueous solution, VOLUSOL®
Trichloroacetic acid (TCA) 500 g/l in aqueous solution, VOLUSOL®
Catalog # 6130.2500
Supplier:  VWR Collection
CAS Number:  
undefined
Trichloroacetic acid (TCA) 500 g/l in aqueous solution, VOLUSOL®
Catalog # 6130.2500
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :163.39 g/mol
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:≤1.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-927-2
UN:2564
ADR:8,II
 :GHS05
 :GHS07
 :GHS09
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177