Pesticide reference standard, Benfuracarb 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10475000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:82560-54-1
- Description:Benfuracarb 10 µg/ml in cyclohexane
- Formula:C₂₀H₃₀N₂O₅S
- Description:Benfuracarb