Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Streptavidin

Streptavidin

Streptavidin
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 189730-1
  • 189730-10
  • Protein/peptide name:
  • Streptavidin
  • Streptavidin
  • CAS No.:
  • 9013-20-1
  • 9013-20-1
  • MDL No.:
  • MFCD00082035
  • MFCD00082035
  • Storage conditions:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 182,00 €
    182,00 €
    1 * 1 mg
       182,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    189730-1
    Protein/peptide name
    Streptavidin
    CAS No.
    9013-20-1
    MDL No.
    MFCD00082035
    Storage conditions
    Freezer
  • Plastic bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 717,00 €
    717,00 €
    1 * 10 mg
       717,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    189730-10
    Protein/peptide name
    Streptavidin
    CAS No.
    9013-20-1
    MDL No.
    MFCD00082035
    Storage conditions
    Freezer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.